
MJN110

Synonym(s):
2,5-dioxopyrrolidin-1-yl 4-(bis(4-chlorophenyl)methyl)piperazine-1-carboxylate, Cravatt Reagent
Empirical Formula (Hill Notation):
C22H21Cl2N3O4
CAS Number:
Molecular Weight:
462.33
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing