
Miconazole

Synonym(s):
(±)-1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, Miconazole
Empirical Formula (Hill Notation):
C18H14Cl4N2O
CAS Number:
Molecular Weight:
416.13
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3