Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo redox buffer
Products
Product Category
buffers (3)
Manufacturer
Physical Form
Volume (ml)
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
MT51350060
Mettler-Toledo redox buffer,
volume 250 mL, 220 mV/pH 7
Expand
View Pricing
MT51350064
Mettler-Toledo redox buffer,
volume 6 x 30 mL, 468 mV/pH 0.1
Expand
View Pricing
MT51350062
Mettler-Toledo redox buffer,
volume 6 x 250 mL, 220 mV/pH 7
Expand
View Pricing