
Methyl trans-4-oxo-2-pentenoate

Synonym(s):
Methyl trans-β-acetylacrylate
Linear Formula:
CH3COCH=CHCO2CH3
CAS Number:
Molecular Weight:
128.13
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing