
Metaraminol tartrate

Synonym(s):
Metaraminol (+)-bitartrate salt, (−)-m-Hydroxyphenylpropanolamine bitartrate salt
Empirical Formula (Hill Notation):
C9H13NO2 · C4H6O6
CAS Number:
Molecular Weight:
317.29
Products
Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing