
Manganese

Synonym(s):
Manganese
Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Element
Quality Segment
Showing 1-20 of 57
Page 1 of 3
Compare
Product Number
Description
Pricing
Page 1 of 3