Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC)
Products
Brand
Sigma-Aldrich (8)
Color
Physical Form
Chemical Composition
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
LUP-514
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
16:0-20:4 PC 100 %
Expand
View Pricing
LUP-516
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
18:0-18:2 PC 100 %
Expand
View Pricing
LUP-513
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
16:0-18:2 PC 100 %
Expand
View Pricing
LUP-515
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
18:0-18:1 PC 100 %
Expand
View Pricing
LUP-519
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
18:1-16:0 PC 100 %
Expand
View Pricing
LUP-517
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
18:0-20:4 PC 100 %
Expand
View Pricing
LUP-518
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
18:1-14:0 PC 100 %
Expand
View Pricing
LUP-520
Lyophilized Liposomes - Unsaturated Phosphatidylcholine (PC),
18:1-18:0 PC 100 %
Expand
View Pricing