
(-)-Quinine

Synonym(s):
(-)-Quinine, Quinine, 6′-Methoxycinchonidine
Empirical Formula (Hill Notation):
C20H24N2O2
CAS Number:
Molecular Weight:
324.42
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing