
Lorazepam Related Compound E

Synonym(s):
6-Chloro-4-(2-chlorophenyl)-2-quinazolinemethanol
Empirical Formula (Hill Notation):
C15H10Cl2N2O
CAS Number:
Molecular Weight:
305.16
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2