
Loratadine Impurity F

Synonym(s):
Ethyl 4-[(11RS)-8-chloro-11-fluoro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl]piperidine-1-carboxylate
Empirical Formula (Hill Notation):
C22H23ClFN2O2
CAS Number:
Molecular Weight:
401.88
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2