
Leucomalachite Green-d5

Synonym(s):
Bis-(4-dimethylaminophenyl)phenyl-d5-methane, LMG-D5
Linear Formula:
C6D5CH[C6H4N(CH3)2]2
CAS Number:
Molecular Weight:
335.50
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing