
(-)-Bilobalide

Synonym(s):
Bilobalide
Empirical Formula (Hill Notation):
C15H18O8
CAS Number:
Molecular Weight:
326.30
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing