Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Krypton-
78
Kr
Linear Formula:
78
Kr
CAS Number:
33580-79-9
Molecular Weight:
77.92
Products
Product Category
gases (6)
stable isotopes (6)
Brand
Sigma-Aldrich (6)
Formula Weight
Physical Form
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
601829
Krypton-
78
Kr,
8 atom %
Expand
View Pricing
601853
Krypton-
78
Kr,
99 atom %
Expand
View Pricing
601802
Krypton-
78
Kr,
1-3 atom %
Expand
View Pricing
601837
Krypton-
78
Kr,
50 atom %
Expand
View Pricing
601845
Krypton-
78
Kr,
90 atom %
Expand
View Pricing
601810
Krypton-
78
Kr,
4-5 atom %
Expand
View Pricing