
Itraconazole

Synonym(s):
Oriconazole, R51211, Sporanox, Itraconazole
Empirical Formula (Hill Notation):
C35H38Cl2N8O4
CAS Number:
Molecular Weight:
705.63
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4