
Ionomycin from Streptomyces conglobatus

Empirical Formula (Hill Notation):
C41H72O9
CAS Number:
Molecular Weight:
709.00
Products
Product Category
Brand
Biological Source
Formula Weight
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing