
Human Fibronectin

Synonym(s):
Fibronectin
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Product Line
Purity
Recombinant Host
Tissue
Quality Segment
Sterility
Technique
Showing 1-11 of 11
Compare
Product Number
Description
Pricing