
Hemin

Synonym(s):
Chloro(protoporphyrinato)iron(III), Chlorohemin, Chloroprotoporphyrin IX iron(III), Ferriprotoporphyrin IX chloride, Hemin(chloride)
Empirical Formula (Hill Notation):
C34H32ClFeN4O4
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing