
Guanidine hydrochloride solution

Synonym(s):
Aminoformamidine hydrochloride, Guanidinium chloride, Guanidium chloride
Empirical Formula (Hill Notation):
CH5N3 · HCl
CAS Number:
Molecular Weight:
95.53
Products
Product Category
Brand
Feature
Formula Weight
Gene Alias
pH Value
Physical Form
Product Line
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing