
Glutardialdehyde solution 25%

Synonym(s):
Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial
Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Products
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing