
Drospirenone

Synonym(s):
6β,7β:15β,16β -Dimethylene-3-oxo-17α-pregn-4-ene-21,17-carbolactone, Dihydrospirorenone
Empirical Formula (Hill Notation):
C24H30O3
CAS Number:
Molecular Weight:
366.49
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4