
Domperidone

Synonym(s):
4-(5-Chloro-2-oxo-1-benzimidazolinyl)-1-[3-(2-oxobenzimidazolinyl)propyl]piperidine
Empirical Formula (Hill Notation):
C22H24ClN5O2
CAS Number:
Molecular Weight:
425.91
Products
Product Category
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing