Skip to Content
Merck
Diethyltoluamide Related Compound A certified reference material, pharmaceutical secondary standard

Diethyltoluamide Related Compound A

Diethyltoluamide Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
N,N-Diethyl-4-methylbenzamide, N,N-Diethyl-4-toluamide, p-DEET, NSC 5686
Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27

Products

Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade