
Diethyl methylphosphonite

Synonym(s):
Diethoxymethylphosphine
Empirical Formula (Hill Notation):
C5H13O2P
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing