
Diacerein

Synonym(s):
1,8-Diacetoxy-3-carboxyanthraquinone
Empirical Formula (Hill Notation):
C19H12O8
CAS Number:
Molecular Weight:
368.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing