Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Devarda′s alloy
Synonym(s):
Devarda′s alloy
Linear Formula:
AlCuZn
CAS Number:
8049-11-4
Products
Brand
Sigma-Aldrich (2)
Supelco (2)
Melting Point (°C)
Physical Form
Product Line
Chemical Composition
Quality Segment
Application
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.05341
Devarda′s alloy,
for analysis EMSURE
®
Expand
View Pricing
483907
Devarda′s alloy,
powder, −100 mesh
Expand
View Pricing
269484
Devarda′s alloy,
filings, +100 mesh
Expand
View Pricing
31385
Devarda′s alloy,
puriss. p.a.
Expand
View Pricing