
(+)-β-Pinene

Synonym(s):
(1R,5R)-2(10)-Pinene, (1R,5R)-6,6-Dimethyl-2-methylenebicyclo[3.1.1]heptane
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2