![[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/227/014/768064c2-9ae6-47bd-8550-e2aa5c3b61de/640/768064c2-9ae6-47bd-8550-e2aa5c3b61de.png)
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt
![[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/227/014/768064c2-9ae6-47bd-8550-e2aa5c3b61de/640/768064c2-9ae6-47bd-8550-e2aa5c3b61de.png)
Synonym(s):
DAGO, DAMGO, Tyr-D-Ala-Gly-N-methyl-Phe-Gly-ol
Empirical Formula (Hill Notation):
C26H35N5O6
CAS Number:
Molecular Weight:
513.59
Products
Product Category
Brand
Formula Weight
Gene Alias
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing