
Clindamycin hydrochloride

Synonym(s):
Clindamycin hydrochloride monohydrate, (7S)-7-Chloro-7-deoxylincomycin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C18H33ClN2O5S · HCl · H2O
CAS Number:
Molecular Weight:
479.46
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2