
Citalopram Related Compound G

Synonym(s):
5-Chloro-1-(4-fluorophenyl)-1,3-dihydro-N,N-dimethyl-1-isobenzofuranpropanamine hydrobromide
Empirical Formula (Hill Notation):
C19H21ClFNO · HBr
CAS Number:
Molecular Weight:
414.74
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2