
cis-4,7,10,13,16,19-Docosahexaenoic acid

Synonym(s):
DHA
Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing