
Chlorfluazuron

Empirical Formula (Hill Notation):
C20H9Cl3F5N3O3
CAS Number:
Molecular Weight:
540.65
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2