
Chamazulene

Synonym(s):
7-Ethyl-1,4-dimethylazulene, Chamazulen, Dimethulene
Empirical Formula (Hill Notation):
C14H16
CAS Number:
Molecular Weight:
184.28
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing