
Cetirizine impurity standard

Synonym(s):
Cetirizine dihydrochloride, [2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid dihydrochloride
Empirical Formula (Hill Notation):
C21H25ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
461.81
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing