
Cerium(IV) oxide-yttria doped

Synonym(s):
Cerium yttrium oxide, YDC, Yttria doped ceria, Yttrium doped ceria
Empirical Formula (Hill Notation):
CeO2/Y2O3
CAS Number:
Products
Brand
Sustainability
Feature
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing