
Cannabidiolic acid

Synonym(s):
CBDA
Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing