
Calcitriol

Synonym(s):
1α,25-Dihydroxyvitamin D3, 1α,25-Dihydroxycholecalciferol, Calcitriol
Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Products
Product Category
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Tissue
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing