
Buspirone hydrochloride

Synonym(s):
N-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride, Buspirone hydrochloride
Empirical Formula (Hill Notation):
C21H31N5O2 · HCl
CAS Number:
Molecular Weight:
421.96
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing