
Budesonide

Synonym(s):
16,17-Butylidenebis(oxy)-11,21-dihydroxypregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing