
Benzethonium chloride

Synonym(s):
(Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Phemerol chloride, Benzethonium chloride
Empirical Formula (Hill Notation):
C27H42ClNO2
CAS Number:
Molecular Weight:
448.08
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing