
Benazepril Related Compound G

Synonym(s):
{3-[1-Ethoxycarbonyl-3-phenyl-(1S)-propyl]amino-2,3,4,5-tetrahydro-2-oxo-1H-1-(3S)-benzazepine}-1-acetic acid ethyl ester
Empirical Formula (Hill Notation):
C26H32N2O5
CAS Number:
Molecular Weight:
452.54
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2