
Atorvastatin Related Compound E

Synonym(s):
(βS,δS)-2-(4-Fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic acid hemicalcium salt, 3S,5S-Atorvastatin calcium
Empirical Formula (Hill Notation):
C66H68CaF2N4O10
CAS Number:
Molecular Weight:
1155.34
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2