
Assay-isotopic standard for silver

Synonym(s):
Silver nitrate, Nitric acid silver(I) salt
Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Isotope
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing