Skip to Content
AZ
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Arsenazo III
Synonym(s):
Arsenazo III, 2,2′-(1,8-Dihydroxy-3,6-disulfonaphthylene-2,7-bisazo)bisbenzenearsonic acid, 2,7-Bis(2-arsonophenylazo)chromotropic acid
Linear Formula:
(HO)
2
C
10
H
2
(SO
3
H)
2
(N=NC
6
H
4
AsO
3
H
2
)
2
CAS Number:
1668-00-4
Molecular Weight:
776.37
EC Number:
216-788-6
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.10107
Arsenazo III,
metal indicator
Expand
View Pricing
A92775
Arsenazo III,
calcium-sensitive dye, Powder
Expand
View Pricing