
Arjunic acid

Synonym(s):
(2α,3β,19α)-2,3,19-Trihydroxyolean-12-en-28-oic acid, Arjuntriterpenic acid
Empirical Formula (Hill Notation):
C30H48O5
CAS Number:
Molecular Weight:
488.70
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing