
Anti-KLK3 antibody produced in rabbit

Synonym(s):
Anti-γ-Seminoprotein antibody produced in rabbit, Anti-Kallikrein-3 antibody produced in rabbit, Anti-P-30 antigen antibody produced in rabbit, Anti-PSA antibody produced in rabbit, Anti-Prostate-specific antigen precursor antibody produced in rabbit, Anti-Semenogelase antibody produced in rabbit, Anti-Seminin antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing