![Anti-CARS antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/201/605/d7ad3a45-0be2-47fe-bfec-d2858408cb2e/640/d7ad3a45-0be2-47fe-bfec-d2858408cb2e.jpg)
Anti-CARS antibody produced in rabbit
![Anti-CARS antibody produced in rabbit Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/201/605/d7ad3a45-0be2-47fe-bfec-d2858408cb2e/640/d7ad3a45-0be2-47fe-bfec-d2858408cb2e.jpg)
Synonym(s):
Anti-CysRS antibody produced in rabbit, Anti-Cysteine--tRNA ligase antibody produced in rabbit, Anti-Cysteinyl-tRNA synthetase, cytoplasmic antibody produced in rabbit, Anti-CYSRS, Anti-Cysteinyl-tRNA synthetase, Anti-MGC:11246
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing