
Amphotericin B for microbiological assay

Synonym(s):
Amphotericin B
Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing