
Ammonium cerium(IV) sulfate dihydrate

Synonym(s):
Ceric ammonium sulfate, Cerium(IV) ammonium sulfate, Ammonium cerium(IV) sulfate dihydrate
Linear Formula:
(NH4)4Ce(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-4 of 4