
Ammonium-15N chloride

Synonym(s):
15N Labeled ammonium chloride
Linear Formula:
15NH4Cl
CAS Number:
Molecular Weight:
54.48
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Application
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing