
Aminolevulinic acid hydrochloride

Synonym(s):
5-Aminolevulinic acid hydrochloride, δ-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, 5-Aminolaevulinic acid hydrochloride, ALA
Linear Formula:
NH2CH2C(O)CH2CH2COOH · HCl
CAS Number:
Molecular Weight:
167.59
Products
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2